Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135457
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281769.1
NP_001268698.1
3
429
NM_001281771.2
NP_001268700.1
3
429
NM_001281772.2
NP_001268701.1
5
628
NM_001281773.2
NP_001268702.1
4
500
NM_001281774.2
NP_001268703.1
4
500
NM_001281775.2
NP_001268704.1
3
429
NM_001281776.2
NP_001268705.1
3
429
NM_001281777.2
NP_001268706.1
3
429
NM_001281778.2
NP_001268707.1
3
429
NM_001281779.2
NP_001268708.1
3
429
NM_001281780.2
NP_001268709.1
3
429
NM_001281781.2
NP_001268710.1
3
429
NM_001281782.2
NP_001268711.1
3
429
NM_001281783.2
NP_001268712.1
3
429
NM_001281784.2
NP_001268713.1
3
429
NM_012408.5
NP_036540.3
3
429
NM_183047.3
NP_898868.1
3
429
NM_183048.3
NP_898869.1
3
429
XM_005260356.4
XP_005260413.1
3
466
XM_005260358.2
XP_005260415.1
2
208
XM_005260360.4
XP_005260417.1
3
931
XM_005260362.2
XP_005260419.1
3
281
XM_005260364.2
XP_005260421.1
3
266
XM_005260366.2
XP_005260423.1
3
222
XM_006723760.3
XP_006723823.1
3
604
XM_006723762.3
XP_006723825.1
3
305
XM_006723763.1
XP_006723826.1
3
295
XM_006723766.1
XP_006723829.1
3
239
XM_011528749.2
XP_011527051.1
3
1110
XM_011528751.1
XP_011527053.1
3
214
XM_011528752.1
XP_011527054.1
2
196
XM_011528753.1
XP_011527055.1
4
300
XM_011528754.1
XP_011527056.1
3
227
XM_017027760.1
XP_016883249.1
2
211
XM_017027761.1
XP_016883250.1
3
677
XM_017027762.1
XP_016883251.1
5
1519
XM_017027763.1
XP_016883252.1
3
357
XM_017027764.1
XP_016883253.1
3
876
XM_017027765.1
XP_016883254.1
3
281
XM_017027766.1
XP_016883255.1
3
281
XM_017027767.1
XP_016883256.1
3
1021
XM_017027768.1
XP_016883257.1
3
281
XM_017027769.1
XP_016883258.1
3
266
XM_017027770.1
XP_016883259.1
4
300
XM_017027771.1
XP_016883260.1
3
266
XM_017027772.1
XP_016883261.1
3
239
XM_017027773.1
XP_016883262.1
3
281
XM_017027774.1
XP_016883263.1
2
184
XM_017027775.1
XP_016883264.1
3
227
XM_017027776.1
XP_016883265.1
3
266
XM_017027777.1
XP_016883266.1
2
184
XM_017027778.1
XP_016883267.1
2
196
XM_017027779.1
XP_016883268.1
2
196
XM_017027780.1
XP_016883269.1
2
211
GenBank mRNA
AB032951.1
-
3
404
AF454056.1
-
3
247
AK000275.1
-
3
246
AK122966.1
-
3
239
AK225857.1
-
3
237
AK294511.1
-
5
438
AK295747.1
-
3
313
AK299899.1
-
4
312
AK309228.1
-
3
298
AL137703.1
-
2
241
BC092432.1
-
3
269
BC136608.1
-
3
260
9020598
BC144289.1
-
3
260
9052809
BC146802.1
-
3
270
8860316
BI553958.1
-
3
275
BX641005.1
-
4
445
DQ082998.1
-
2
210
DQ082999.1
-
2
210
DQ368669.1
-
2
210
DQ368670.1
-
2
210
DQ368671.1
-
2
210
DQ368672.1
-
2
210
DQ368673.1
-
2
210
DQ368674.1
-
2
210
DQ368675.1
-
2
210
DQ368676.1
-
2
210
DQ368677.1
-
2
210
DQ368678.1
-
2
210
DQ368679.1
-
2
210
DQ368680.1
-
2
210
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0