Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
ZMYND8
Gene Name:
zinc finger MYND-type containing 8
Gene Aliases:
PRKCBP1, PRO2893, RACK7
Chromosome Location:
Chr.20: 47209214 - 47357780 on Build GRCh38
UniGene ID:
Hs.446240
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135459
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001281771.2
NP_001268700.1
21
4352
NM_001281772.2
NP_001268701.1
24
4932
NM_001281773.2
NP_001268702.1
24
4963
NM_001281774.2
NP_001268703.1
24
4825
NM_001281775.2
NP_001268704.1
23
4892
NM_001281776.2
NP_001268705.1
22
4511
NM_001281777.2
NP_001268706.1
23
4679
NM_001281778.2
NP_001268707.1
22
4733
NM_001281779.2
NP_001268708.1
24
4885
NM_001281780.2
NP_001268709.1
23
4801
NM_001281781.2
NP_001268710.1
21
4454
NM_001281782.2
NP_001268711.1
22
4598
NM_001281783.2
NP_001268712.1
22
4808
NM_001281784.2
NP_001268713.1
21
4592
NM_012408.5
NP_036540.3
22
4670
NM_183047.3
NP_898868.1
23
4754
NM_183048.3
NP_898869.1
22
4595
XM_005260356.4
XP_005260413.1
23
4929
XM_005260358.2
XP_005260415.1
22
4671
XM_005260360.4
XP_005260417.1
23
5394
XM_005260362.2
XP_005260419.1
23
4669
XM_005260364.2
XP_005260421.1
23
4654
XM_005260366.2
XP_005260423.1
22
4601
XM_006723760.3
XP_006723823.1
23
5067
XM_006723762.3
XP_006723825.1
23
4768
XM_006723763.1
XP_006723826.1
23
4683
XM_006723766.1
XP_006723829.1
23
4627
XM_011528749.2
XP_011527051.1
23
5573
XM_011528751.1
XP_011527053.1
23
4677
XM_011528752.1
XP_011527054.1
21
4518
XM_011528753.1
XP_011527055.1
24
4688
XM_011528754.1
XP_011527056.1
23
4615
XM_017027760.1
XP_016883249.1
20
4523
XM_017027761.1
XP_016883250.1
23
5140
XM_017027762.1
XP_016883251.1
25
5907
XM_017027763.1
XP_016883252.1
22
4736
XM_017027764.1
XP_016883253.1
21
5188
XM_017027765.1
XP_016883254.1
21
4518
XM_017027766.1
XP_016883255.1
23
4669
XM_017027767.1
XP_016883256.1
22
5400
XM_017027768.1
XP_016883257.1
22
4660
XM_017027769.1
XP_016883258.1
21
4503
XM_017027770.1
XP_016883259.1
23
4604
XM_017027771.1
XP_016883260.1
22
4570
XM_017027772.1
XP_016883261.1
22
4543
XM_017027773.1
XP_016883262.1
21
4380
XM_017027774.1
XP_016883263.1
22
4647
XM_017027775.1
XP_016883264.1
23
4477
XM_017027776.1
XP_016883265.1
20
4362
XM_017027777.1
XP_016883266.1
21
4563
XM_017027779.1
XP_016883268.1
20
3768
XM_017027780.1
XP_016883269.1
19
3699
GenBank mRNA
AF233453.1
-
11
2871
BC146802.1
-
15
3291
8860316
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
positive regulation of filopodium assembly
positive regulation of dendritic spine development
modulation of excitatory postsynaptic potential
regulation of postsynaptic density protein 95 clustering
positive regulation of dendritic spine maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription corepressor activity
chromatin binding
transcription cofactor activity
protein binding
zinc ion binding
histone binding
protein N-terminus binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline