Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165881.2 | NP_001159353.1 | 6 | 2156 | ||
NM_001165882.2 | NP_001159354.1 | 6 | 2163 | |||
NM_001165883.1 | NP_001159355.1 | 4 | 2057 | |||
NM_001165884.2 | NP_001159356.2 | 3 | 1856 | |||
NM_001165885.1 | NP_001159357.1 | 5 | 2153 | |||
NM_001165886.1 | NP_001159358.1 | 4 | 1952 | |||
NM_001165887.1 | NP_001159359.1 | 6 | 2185 | |||
NM_003415.2 | NP_003406.1 | 6 | 2280 | |||
NM_152943.2 | NP_694422.2 | 7 | 2386 | |||
GenBank mRNA | AF317549.1 | - | 6 | 2280 | ||
AF385187.1 | - | 7 | 2236 | |||
AF432217.1 | - | 3 | 1756 | |||
AK027712.1 | - | 7 | 2300 | |||
AY583691.1 | - | 8 | 2132 | |||
BC110542.2 | - | 6 | 2156 | 40036577 | ||
X78926.1 | - | 1 | 306 |