Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRRM1 SIRNA Assays ›
Gene Symbol:
SRRM1
Gene Name:
serine and arginine repetitive matrix 1
Gene Aliases:
160-KD, POP101, SRM160
Chromosome Location:
Chr.1: 24643012 - 24673281 on Build GRCh38
UniGene ID:
Hs.18192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135502
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303448.1
NP_001290377.1
3
402
NM_001303449.1
NP_001290378.1
3
331
NM_005839.3
NP_005830.2
3
402
XM_005245717.2
XP_005245774.1
3
312
XM_005245718.2
XP_005245775.1
3
312
XM_005245720.2
XP_005245777.1
3
312
XM_005245721.2
XP_005245778.1
3
312
XM_005245722.2
XP_005245779.1
3
312
XM_011540478.2
XP_011538780.1
3
326
XM_011540479.1
XP_011538781.1
3
493
XM_011540480.1
XP_011538782.1
3
493
XM_011540481.1
XP_011538783.1
3
493
XM_011540482.1
XP_011538784.1
3
493
XM_011540483.1
XP_011538785.1
3
493
XM_017000011.1
XP_016855500.1
3
326
XM_017000012.1
XP_016855501.1
3
326
XM_017000013.1
XP_016855502.1
3
326
XM_017000014.1
XP_016855503.1
3
326
XM_017000015.1
XP_016855504.1
3
326
XM_017000016.1
XP_016855505.1
3
348
XM_017000017.1
XP_016855506.1
3
326
XM_017000018.1
XP_016855507.1
3
312
XM_017000019.1
XP_016855508.1
3
312
XM_017000020.1
XP_016855509.1
3
312
XM_017000021.1
XP_016855510.1
3
312
XM_017000022.1
XP_016855511.1
3
312
XM_017000023.1
XP_016855512.1
3
493
XM_017000024.1
XP_016855513.1
3
493
XM_017000025.1
XP_016855514.1
3
312
XM_017000026.1
XP_016855515.1
3
312
XM_017000027.1
XP_016855516.1
3
493
XM_017000028.1
XP_016855517.1
3
493
XM_017000029.1
XP_016855518.1
3
493
XM_017000030.1
XP_016855519.1
3
493
XM_017000031.1
XP_016855520.1
3
493
XM_017000032.1
XP_016855521.1
3
493
XM_017000033.1
XP_016855522.1
3
493
XM_017000034.1
XP_016855523.1
3
326
GenBank mRNA
AF048977.1
-
3
183
AF419855.1
-
3
405
AK302498.1
-
3
221
AK307419.1
-
3
331
BC036187.1
-
3
230
4827738
BG720378.1
-
3
405
DC353932.1
-
3
311
KU178512.1
-
3
178
KU178513.1
-
3
178
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
DNA binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline