Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005850.4 | NP_005841.1 | 3 | 1019 | ||
GenBank mRNA | AK097315.1 | - | 3 | 1111 | ||
AK223322.1 | - | 3 | 535 | |||
AK310495.1 | - | 3 | 535 | |||
BC004273.1 | - | 3 | 534 | 3615080 | ||
BC013886.2 | - | 3 | 534 | 3829990 | ||
BC090883.1 | - | 4 | 1139 | 30386953 | ||
BM460550.1 | - | 3 | 577 |