Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005858.3 | NP_005849.1 | 14 | 2606 | ||
XM_011527624.2 | XP_011525926.1 | 14 | 2699 | |||
XM_017026141.1 | XP_016881630.1 | 14 | 2751 | |||
GenBank mRNA | AL050160.1 | - | 8 | 1534 | ||
Y11997.1 | - | 14 | 2597 |