Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047160.2 | NP_001040625.1 | 12 | 3248 | ||
NM_005863.4 | NP_005854.2 | 10 | 3159 | |||
NR_073040.1 | 10 | 3192 | ||||
GenBank mRNA | BC010285.1 | - | 10 | 3091 | 3048980 | |
BX537509.1 | - | 6 | 5797 |