Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277174.1 | NP_001264103.1 | 6 | 2335 | ||
NM_005864.3 | NP_005855.1 | 6 | 2842 | |||
NM_032459.2 | NP_115835.1 | 5 | 2563 | |||
XM_005267256.1 | XP_005267313.1 | 5 | 2333 | |||
GenBank mRNA | AB001466.1 | - | 6 | 2842 | ||
AB001467.1 | - | 5 | 2563 | |||
BC034246.1 | - | 5 | 2111 | 4214788 | ||
BC046403.1 | - | 5 | 2682 |