Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005867.3 | NP_005858.1 | 3 | 443 | ||
GenBank mRNA | AB000099.1 | - | 3 | 443 | ||
BC069729.1 | - | 3 | 405 | 7262631 | ||
BC096162.1 | - | 3 | 442 | 40003276 | ||
BC096163.1 | - | 3 | 442 | 40003277 | ||
BC096164.1 | - | 3 | 442 | 40003278 | ||
DQ179113.1 | - | 3 | 393 |