Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 15, 16 | 2431 | ||
GenBank mRNA | AK098463.1 | - | 8, 9 | 1197 | ||
AK303273.1 | - | 16, 17 | 2356 | |||
BC001976.1 | - | 15, 16 | 2371 | 3459964 | ||
BC007684.2 | - | 15, 16 | 2398 | 3634035 | ||
CR456575.1 | - | 15, 16 | 2374 | |||
X85237.1 | - | 15, 16 | 2373 |