Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006059.3 | NP_006050.3 | 22 | 3826 | ||
XM_006716921.2 | XP_006716984.1 | 22 | 3769 | |||
XM_011518121.1 | XP_011516423.1 | 22 | 3769 | |||
GenBank mRNA | AB208887.1 | - | 21 | 3142 | ||
AF041835.1 | - | 22 | 3825 | |||
BX327938.1 | - | 6 | 738 |