Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IKZF1 SIRNA Assays ›
Gene Symbol:
IKZF1
Gene Name:
IKAROS family zinc finger 1
Gene Aliases:
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
Chromosome Location:
Chr.7: 50303453 - 50405101 on Build GRCh38
UniGene ID:
Hs.435949
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135569
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IKZF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220767.2
NP_001207696.1
4
397
NM_001291838.1
NP_001278767.1
5
624
NM_001291841.1
NP_001278770.1
3
229
NM_001291842.1
NP_001278771.1
3
229
NM_006060.5
NP_006051.1
6
912
XM_011515058.2
XP_011513360.1
7
802
XM_011515059.2
XP_011513361.1
7
804
XM_011515060.2
XP_011513362.1
7
3952
XM_011515061.2
XP_011513363.1
6
740
XM_011515062.2
XP_011513364.1
7
1082
XM_011515063.2
XP_011513365.1
7
999
XM_011515064.2
XP_011513366.1
7
4630
XM_011515065.2
XP_011513367.1
7
767
XM_011515066.2
XP_011513368.1
7
871
XM_011515068.2
XP_011513370.1
6
939
XM_011515069.2
XP_011513371.1
6
4567
XM_011515071.2
XP_011513373.1
6
541
XM_011515072.2
XP_011513374.1
5
479
XM_011515073.2
XP_011513375.1
6
821
XM_011515074.2
XP_011513376.1
5
482
XM_011515075.2
XP_011513377.1
5
761
XM_011515076.2
XP_011513378.1
5
705
XM_017011667.1
XP_016867156.1
6
738
XM_017011668.1
XP_016867157.1
5
445
XM_017011669.1
XP_016867158.1
6
707
XM_017011670.1
XP_016867159.1
6
1068
GenBank mRNA
AK303586.1
-
5
583
U40462.1
-
6
812
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
mesoderm development
covalent chromatin modification
lymphocyte differentiation
erythrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0