Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260641.3 | XP_005260698.1 | 6 | 1431 | ||
XM_005260643.3 | XP_005260700.1 | 5 | 1098 | |||
XM_005260644.3 | XP_005260701.1 | 4 | 781 | |||
XM_017027577.1 | XP_016883066.1 | 6 | 1894 | |||
XM_017027581.1 | XP_016883070.1 | 7 | 1739 | |||
XM_017027582.1 | XP_016883071.1 | 7 | 1779 | |||
XM_017027585.1 | XP_016883074.1 | 5 | 1531 | |||
GenBank mRNA | BC025286.1 | - | 4 | 771 | ||
BC075835.1 | - | 4 | 771 | |||
BC131747.1 | - | 4 | 755 | |||
Y10376.1 | - | 6 | 1410 |