Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006097.4 | NP_006088.2 | 4 | 616 | ||
NM_181526.2 | NP_852667.1 | 3 | 454 | |||
GenBank mRNA | AF176042.1 | - | 3 | 552 | ||
AF176043.1 | - | 2 | 390 | |||
AK130879.1 | - | 4 | 581 | |||
BC002648.2 | - | 3 | 393 | 3608316 | ||
BQ953325.1 | - | 606 | ||||
D82057.1 | - | 4 | 584 |