Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006098.4 | NP_006089.1 | 6 | 792 | ||
GenBank mRNA | AK095666.1 | - | 6 | 664 | ||
AK222488.1 | - | 6 | 792 | |||
AK301468.1 | - | 6 | 718 | |||
AY336089.1 | - | 6 | 693 | |||
BC000214.1 | - | 6 | 810 | 2959178 | ||
BC000366.2 | - | 6 | 780 | 2819249 | ||
BC010119.2 | - | 6 | 767 | 3835636 | ||
BC014256.1 | - | 6 | 769 | 4765908 | ||
BC014788.1 | - | 6 | 776 | 3455986 | ||
BC017287.2 | - | 6 | 757 | 4892271 | ||
BC019093.2 | - | 6 | 770 | 4893346 | ||
BC019362.1 | - | 6 | 774 | 2823761 | ||
BC021993.1 | - | 6 | 792 | 4083071 | ||
BC029996.1 | - | 6 | 619 | |||
BC032006.1 | - | 6 | 771 | 4750184 | ||
BC035460.1 | - | 6 | 773 | |||
CR456978.1 | - | 6 | 686 | |||
CR541909.1 | - | 6 | 686 | |||
GU727632.1 | - | 6 | 686 | |||
M24194.1 | - | 6 | 781 |