Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039619.2 | NP_001034708.1 | 9, 10 | 1334 | ||
NM_001282953.1 | NP_001269882.1 | 8, 9 | 1202 | |||
NM_001282954.1 | NP_001269883.1 | 8, 9 | 1215 | |||
NM_001282955.1 | NP_001269884.1 | 8, 9 | 1117 | |||
NM_001282956.1 | NP_001269885.1 | 5, 6 | 872 | |||
NM_006109.4 | NP_006100.2 | 9, 10 | 1249 | |||
XM_017020927.1 | XP_016876416.1 | 8, 9 | 973 | |||
GenBank mRNA | AB451246.1 | - | 9, 10 | 1025 | ||
AB451370.1 | - | 9, 10 | 1025 | |||
AF015913.1 | - | 9, 10 | 1025 | |||
AF167572.1 | - | 9, 10 | 1116 | |||
AK027737.1 | - | 738 | ||||
AK075251.1 | - | 9, 10 | 1127 | |||
AK300863.1 | - | 8, 9 | 999 | |||
AK301812.1 | - | 8, 9 | 1008 | |||
AK302240.1 | - | 8, 9 | 897 | |||
AK315500.1 | - | 9, 10 | 1065 | |||
AL833612.1 | - | 9, 10 | 1034 | |||
BC025979.2 | - | 9, 10 | 1037 | 3833019 | ||
CR456741.1 | - | 9, 10 | 1025 |