Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039619.2 | NP_001034708.1 | 17 | 2411 | ||
NM_001282953.1 | NP_001269882.1 | 16 | 2279 | |||
NM_001282954.1 | NP_001269883.1 | 16 | 2292 | |||
NM_001282955.1 | NP_001269884.1 | 16 | 2194 | |||
NM_001282956.1 | NP_001269885.1 | 13 | 1949 | |||
NM_006109.4 | NP_006100.2 | 17 | 2326 | |||
XM_017020927.1 | XP_016876416.1 | 16 | 2050 | |||
GenBank mRNA | AF167572.1 | - | 17 | 2195 | ||
AK027737.1 | - | 1465 | ||||
AK075251.1 | - | 17 | 2204 | |||
AK301812.1 | - | 16 | 2085 | |||
AL833612.1 | - | 17 | 2111 | |||
BC025979.2 | - | 17 | 2114 | 3833019 |