Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166010.1 | NP_001159482.1 | 3 | 273 | ||
NM_006117.2 | NP_006108.2 | 3 | 261 | |||
NM_206836.2 | NP_996667.2 | 3 | 271 | |||
NR_028588.1 | 3 | 261 | ||||
XM_006714957.1 | XP_006715020.1 | 3 | 264 | |||
XM_017010170.1 | XP_016865659.1 | 3 | 255 | |||
GenBank mRNA | AB209917.1 | - | 2 | 255 | ||
AF069301.1 | - | 3 | 221 | |||
AF153612.1 | - | 3 | 231 | |||
AF244138.1 | - | 3 | 226 | |||
AF257175.1 | - | 3 | 223 | |||
AK057175.1 | - | 3 | 264 | |||
AK075108.1 | - | 3 | 239 | |||
AK222617.1 | - | 3 | 247 | |||
AK223003.1 | - | 3 | 249 | |||
AK297049.1 | - | 3 | 239 | |||
AL136642.1 | - | 3 | 248 | |||
AU100345.1 | - | 3 | 261 | |||
AY297537.1 | - | 2 | 176 | |||
BC002668.2 | - | 3 | 248 | 3608151 | ||
BC016781.1 | - | 3 | 240 | 4107308 | ||
BC017474.1 | - | 3 | 225 | 3544703 | ||
BC025287.1 | - | 3 | 224 | |||
BC033841.1 | - | 3 | 231 | 3923244 | ||
BC034702.1 | - | 3 | 273 | 4750108 | ||
CR456768.1 | - | 2 | 144 |