Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317836.1 | NP_001304765.1 | 8 | 877 | ||
NM_006315.5 | NP_006306.2 | 7 | 778 | |||
GenBank mRNA | AK057124.1 | - | 7 | 1715 | ||
AK125801.1 | - | 8 | 1722 | |||
AK225892.1 | - | 7 | 777 | |||
AK300290.1 | - | 8 | 786 | |||
BC107061.2 | - | 5 | 403 | 40006331 | ||
BQ882065.1 | - | 7 | 621 | |||
BX648409.1 | - | 3 | 1126 | |||
CA454695.1 | - | 173 |