Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286277.1 | NP_001273206.1 | 2, 3 | 396 | ||
NM_001286278.1 | NP_001273207.1 | 2, 3 | 396 | |||
NM_001286279.1 | NP_001273208.1 | 2, 3 | 396 | |||
NM_006322.5 | NP_006313.1 | 2, 3 | 396 | |||
XM_005268293.1 | XP_005268350.1 | 2, 3 | 391 | |||
XM_011537462.2 | XP_011535764.1 | 2, 3 | 349 | |||
XM_011537463.2 | XP_011535765.1 | 3, 4 | 325 | |||
XM_011537464.2 | XP_011535766.1 | 2, 3 | 1129 | |||
XM_017020323.1 | XP_016875812.1 | 3, 4 | 743 | |||
GenBank mRNA | AF042378.1 | - | 2, 3 | 275 | ||
AJ003061.1 | - | 2, 3 | 191 | |||
AJ003062.1 | - | 2, 3 | 191 | |||
AK302537.1 | - | 2, 3 | 343 | |||
AK314785.1 | - | 2, 3 | 396 | |||
BC007763.2 | - | 2, 3 | 346 | |||
BC013781.1 | - | 2, 3 | 336 | 3867019 | ||
BC046634.2 | - | 2, 3 | 379 | 5295831 | ||
BT009772.1 | - | 2, 3 | 191 |