Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006328.3 | NP_006319.1 | 2 | 1914 | ||
GenBank mRNA | AB209007.1 | - | 1 | 4614 | ||
AF080561.1 | - | 2 | 1855 | |||
AF315632.1 | - | 2 | 1870 | |||
AK021768.1 | - | 2 | 1866 | |||
AK222830.1 | - | 2 | 1867 | |||
AK297907.1 | - | 2 | 606 | |||
BC000488.2 | - | 2 | 1850 | 2819856 | ||
BC023619.2 | - | 2 | 1850 | |||
KU178520.1 | - | 2 | 1775 |