Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279357.1 | NP_001266286.1 | 5, 6 | 514 | ||
NM_001279358.1 | NP_001266287.1 | 6, 7 | 562 | |||
NM_001279359.1 | NP_001266288.1 | 6, 7 | 522 | |||
NM_001279360.1 | NP_001266289.1 | 6, 7 | 504 | |||
NM_006330.3 | NP_006321.1 | 6, 7 | 562 | |||
XM_005251127.4 | XP_005251184.1 | 6, 7 | 362 | |||
XM_017012956.1 | XP_016868445.1 | 5, 6 | 473 | |||
XM_017012957.1 | XP_016868446.1 | 7, 8 | 512 | |||
XM_017012958.1 | XP_016868447.1 | 6, 7 | 439 | |||
XM_017012959.1 | XP_016868448.1 | 7, 8 | 561 | |||
XM_017012960.1 | XP_016868449.1 | 5, 6 | 379 | |||
GenBank mRNA | AA307642.1 | - | 6, 7 | 320 | ||
AF052112.1 | - | 6, 7 | 562 | |||
AF077198.1 | - | 6, 7 | 373 | |||
AF077199.1 | - | 5, 6 | 325 | |||
AF081281.1 | - | 6, 7 | 403 | |||
AF291053.1 | - | 6, 7 | 386 | |||
AK289400.1 | - | 6, 7 | 446 | |||
AK313739.1 | - | 6, 7 | 399 | |||
BC008652.2 | - | 6, 7 | 450 | 3865775 | ||
BC010397.1 | - | 6, 7 | 374 | 4109335 | ||
CR457103.1 | - | 6, 7 | 368 | |||
DA817192.1 | - | 6, 7 | 346 | |||
DA881616.1 | - | 6, 7 | 394 |