Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199873.1 | NP_001186802.1 | 3 | 861 | ||
NM_001199874.1 | NP_001186803.1 | 3 | 790 | |||
NM_003779.3 | NP_003770.1 | 3 | 840 | |||
XM_005245566.1 | XP_005245623.1 | 4 | 562 | |||
XM_011510093.1 | XP_011508395.1 | 3 | 396 | |||
XM_017002714.1 | XP_016858203.1 | 3 | 417 | |||
XM_017002715.1 | XP_016858204.1 | 3 | 396 | |||
GenBank mRNA | AB024435.1 | - | 1 | 139 | ||
AF038661.1 | - | 3 | 367 | |||
AK023311.1 | - | 3 | 396 | |||
AK056851.1 | - | 2 | 1397 | |||
AK092493.1 | - | 4 | 322 | |||
AK291455.1 | - | 3 | 394 | |||
BC000276.1 | - | 3 | 343 | 3357342 | ||
BC006099.1 | - | 3 | 357 | 3677118 | ||
BC009985.2 | - | 3 | 319 | 4124756 | ||
Y12509.1 | - | 1 | 134 |