Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006342.2 | NP_006333.1 | 8, 9 | 1911 | ||
XM_005247929.2 | XP_005247986.1 | 8, 9 | 1909 | |||
XM_005247930.2 | XP_005247987.1 | 7, 8 | 938 | |||
XM_011513386.2 | XP_011511688.1 | 8, 9 | 1909 | |||
XM_017007653.1 | XP_016863142.1 | 8, 9 | 1963 | |||
GenBank mRNA | AF093543.1 | - | 8, 9 | 1864 | ||
AF479620.1 | - | 7, 8 | 777 | |||
AJ243997.1 | - | 8, 9 | 1856 | |||
AK302463.1 | - | 8, 9 | 1882 | |||
AK304579.1 | - | 7, 8 | 1640 | |||
AK310660.1 | - | 7, 8 | 1634 | |||
BC106071.1 | - | 8, 9 | 1872 | 6148176 | ||
BC111771.1 | - | 8, 9 | 1824 | 40034847 |