Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 3, 4 | 713 | ||
XM_005263565.4 | XP_005263622.1 | 3, 4 | 848 | |||
XM_005263568.4 | XP_005263625.1 | 3, 4 | 848 | |||
XM_011510490.2 | XP_011508792.1 | 3, 4 | 748 | |||
XM_017003164.1 | XP_016858653.1 | 3, 4 | 718 | |||
XM_017003165.1 | XP_016858654.1 | 3, 4 | 765 | |||
GenBank mRNA | AK316584.1 | - | 3, 4 | 720 | ||
AL705756.1 | - | 2, 3 | 173 | |||
BC114917.1 | - | 3, 4 | 427 | 40068588 | ||
BC114918.1 | - | 3, 4 | 427 | 40012335 | ||
CA454601.1 | - | 1, 2 | 103 | |||
CN371299.1 | - | 2, 3 | 348 | |||
U08023.1 | - | 3, 4 | 728 |