Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006345.3 | NP_006336.3 | 9 | 962 | ||
XM_017007654.1 | XP_016863143.1 | 9 | 996 | |||
GenBank mRNA | AF006621.2 | - | 9 | 936 | ||
AK225470.1 | - | 9 | 899 | |||
AK299917.1 | - | 10 | 1056 | |||
AK312838.1 | - | 9 | 962 | |||
AY319413.1 | - | 10 | 841 | |||
AY594282.1 | - | 9 | 816 | |||
BC000240.1 | - | 9 | 924 | |||
BC007732.2 | - | 9 | 923 | 4109702 | ||
BC016949.1 | - | 9 | 941 | 3850521 | ||
BC022981.1 | - | 5 | 353 | |||
KU178526.1 | - | 9 | 816 | |||
KU178527.1 | - | 9 | 816 |