Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161520.1 | NP_001154992.1 | 7 | 1236 | ||
NM_006348.3 | NP_006339.3 | 7 | 1236 | |||
NM_181733.2 | NP_859422.2 | 7 | 1236 | |||
GenBank mRNA | AF058718.1 | - | 7 | 762 | ||
AK314270.1 | - | 7 | 1234 | |||
BC068540.1 | - | 7 | 983 | 4374289 | ||
BI597200.1 | - | 7 | 646 |