Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006364.3 | NP_006355.2 | 16 | 2386 | ||
XM_005267262.1 | XP_005267319.1 | 17 | 2173 | |||
XM_011536355.2 | XP_011534657.1 | 17 | 2173 | |||
XM_017020928.1 | XP_016876417.1 | 16 | 2101 | |||
GenBank mRNA | AK127355.1 | - | 12 | 2465 | ||
AK293944.1 | - | 16 | 1763 | |||
AK299862.1 | - | 15 | 1983 | |||
AK312259.1 | - | 16 | 1968 | |||
BX537566.1 | - | 8 | 873 | |||
X97064.1 | - | 16 | 2027 |