Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006366.2 | NP_006357.1 | 8 | 1058 | ||
XM_011514233.2 | XP_011512535.1 | 7 | 1269 | |||
GenBank mRNA | AK293134.1 | - | 6 | 659 | ||
AK295600.1 | - | 5 | 594 | |||
AK297538.1 | - | 7 | 774 | |||
AK308461.1 | - | 8 | 877 | |||
AK312362.1 | - | 8 | 1186 | |||
AL832154.1 | - | 6 | 720 | |||
BC008481.1 | - | 8 | 851 | 4295984 | ||
CR457022.1 | - | 7 | 815 | |||
U02390.1 | - | 8 | 898 |