Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNCRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159673.1 | NP_001153145.1 | 5 | 539 | ||
NM_001159674.1 | NP_001153146.1 | 6 | 667 | |||
NM_001159675.1 | NP_001153147.1 | 5 | 573 | |||
NM_001159676.1 | NP_001153148.1 | 5 | 573 | |||
NM_001159677.1 | NP_001153149.1 | 6 | 1102 | |||
NM_001253771.1 | NP_001240700.1 | 5 | 481 | |||
NM_006372.4 | NP_006363.4 | 6 | 1102 | |||
XM_005248635.4 | XP_005248692.1 | 6 | 723 | |||
XM_005248636.2 | XP_005248693.1 | 5 | 953 | |||
XM_005248637.2 | XP_005248694.1 | 6 | 686 | |||
XM_017010176.1 | XP_016865665.1 | 6 | 723 | |||
XM_017010177.1 | XP_016865666.1 | 6 | 723 | |||
XM_017010178.1 | XP_016865667.1 | 6 | 683 | |||
XM_017010179.1 | XP_016865668.1 | 6 | 658 | |||
XM_017010180.1 | XP_016865669.1 | 5 | 738 | |||
GenBank mRNA | AB209098.1 | - | 5 | 479 | ||
AF037448.1 | - | 6 | 1086 | |||
AF155568.1 | - | 6 | 765 | |||
AK222776.1 | - | 6 | 711 | |||
AK299796.1 | - | 5 | 534 | |||
AK313572.1 | - | 6 | 722 | |||
AY034481.1 | - | 5 | 561 | |||
AY034482.1 | - | 5 | 561 | |||
AY034483.1 | - | 5 | 561 | |||
BC015575.2 | - | 6 | 643 | |||
BC024283.1 | - | 6 | 670 | |||
BC032643.1 | - | 6 | 658 | 5495201 | ||
BC040844.1 | - | 5 | 481 | 4513184 | ||
CV806885.1 | - | 2 | 103 |