Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142287.1 | NP_001135759.1 | 6 | 802 | ||
NM_006378.3 | NP_006369.3 | 6 | 802 | |||
XM_005251654.3 | XP_005251711.1 | 4 | 636 | |||
XM_011518123.2 | XP_011516425.1 | 8 | 886 | |||
XM_011518124.2 | XP_011516426.1 | 8 | 845 | |||
XM_011518125.1 | XP_011516427.1 | 6 | 773 | |||
XM_011518127.2 | XP_011516429.1 | 7 | 801 | |||
XM_011518128.2 | XP_011516430.1 | 6 | 726 | |||
XM_011518129.1 | XP_011516431.1 | 5 | 655 | |||
XM_011518130.2 | XP_011516432.1 | 5 | 890 | |||
XM_011518131.2 | XP_011516433.1 | 4 | 828 | |||
XM_011518133.2 | XP_011516435.1 | 5 | 752 | |||
XM_011518134.2 | XP_011516436.1 | 3 | 693 | |||
XM_017014193.1 | XP_016869682.1 | 7 | 2496 | |||
XM_017014194.1 | XP_016869683.1 | 7 | 770 | |||
XM_017014195.1 | XP_016869684.1 | 6 | 707 | |||
XM_017014196.1 | XP_016869685.1 | 6 | 1044 | |||
XM_017014197.1 | XP_016869686.1 | 5 | 928 | |||
XM_017014198.1 | XP_016869687.1 | 4 | 865 | |||
XM_017014199.1 | XP_016869688.1 | 8 | 845 | |||
GenBank mRNA | AB210030.1 | - | 4 | 1007 | ||
AK293749.1 | - | 5 | 912 | |||
AK316107.1 | - | 6 | 996 | |||
BC054500.1 | - | 6 | 802 | 5551182 | ||
BX648216.1 | - | 4 | 594 | |||
U60800.1 | - | 2 | 317 |