Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271888.1 | NP_001258817.1 | 5 | 1033 | ||
NM_001271889.1 | NP_001258818.1 | 4 | 921 | |||
NM_001301224.1 | NP_001288153.1 | 5 | 1083 | |||
NM_006383.3 | NP_006374.1 | 6 | 1068 | |||
NR_125435.1 | 5 | 956 | ||||
XM_006720374.2 | XP_006720437.1 | 7 | 893 | |||
GenBank mRNA | AB012955.1 | - | 6 | 754 | ||
BC033108.1 | - | 5 | 1033 | 4841632 | ||
BC047381.1 | - | 6 | 996 | 5287072 | ||
BC057292.1 | - | 6 | 996 | |||
BE907622.1 | - | 313 | ||||
BI523557.1 | - | 3 | 468 | |||
CN371149.1 | - | 2 | 332 |