Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277764.1 | NP_001264693.1 | 5 | 650 | ||
NM_006384.3 | NP_006375.2 | 5 | 530 | |||
NR_102427.1 | 5 | 554 | ||||
NR_102428.1 | 5 | 420 | ||||
XM_006720375.1 | XP_006720438.1 | 5 | 525 | |||
GenBank mRNA | AB451276.1 | - | 5 | 368 | ||
AB451406.1 | - | 5 | 368 | |||
BC000846.2 | - | 5 | 410 | 3457844 | ||
BG759961.1 | - | 5 | 420 | |||
BI520476.1 | - | 5 | 554 | |||
CD622547.1 | - | 5 | 537 | |||
CR456955.1 | - | 5 | 368 | |||
JQ246073.1 | - | 5 | 488 | |||
U82226.1 | - | 5 | 414 | |||
U83236.1 | - | 5 | 436 | |||
U85611.1 | - | 5 | 434 |