Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003793.3 | NP_003784.2 | 3 | 527 | ||
XM_011545328.2 | XP_011543630.1 | 3 | 277 | |||
GenBank mRNA | AF071748.1 | - | 1 | 91 | ||
AF088886.2 | - | 3 | 527 | |||
AF136279.1 | - | 3 | 526 | |||
AJ007331.1 | - | 3 | 482 | |||
AK313657.1 | - | 3 | 495 | |||
BC011682.2 | - | 3 | 521 | 3535532 | ||
BC036451.1 | - | 3 | 502 | 4830274 |