Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHERP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006387.5 | NP_006378.3 | 17 | 4018 | ||
GenBank mRNA | AB209730.1 | - | 8 | 2395 | ||
AF536542.1 | - | 17 | 3985 | |||
BC021294.2 | - | 17 | 4003 | 4053575 | ||
U94836.2 | - | 16 | 3920 |