Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003794.3 | NP_003785.1 | 2, 3 | 313 | ||
XM_017007414.1 | XP_016862903.1 | 2, 3 | 305 | |||
GenBank mRNA | AF065485.1 | - | 2, 3 | 271 | ||
AK001835.1 | - | 2, 3 | 275 | |||
BC018762.1 | - | 2, 3 | 289 | 4641014 | ||
BI669801.1 | - | 2, 3 | 317 | |||
DA289870.1 | - | 2, 3 | 315 |