Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006402.2 | NP_006393.2 | 4 | 752 | ||
GenBank mRNA | AK054769.1 | - | 3 | 1728 | ||
AK056679.1 | - | 1 | 1655 | |||
BC062619.1 | - | 4 | 770 | 5190406 |