Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006408.3 | NP_006399.1 | 6, 7 | 575 | ||
XM_005249581.4 | XP_005249638.1 | 6, 7 | 544 | |||
GenBank mRNA | AF007791.1 | - | 5, 6 | 398 | ||
AF038451.1 | - | 6, 7 | 446 | |||
AF087879.1 | - | 6, 7 | 537 | |||
AF088867.1 | - | 6, 7 | 535 | |||
AF115926.1 | - | 6, 7 | 445 | |||
AK223067.1 | - | 6, 7 | 466 | |||
AK312942.1 | - | 6, 7 | 456 | |||
AY359009.1 | - | 6, 7 | 535 | |||
BC015503.1 | - | 6, 7 | 444 | 3852448 | ||
BT007048.1 | - | 5, 6 | 388 | |||
FJ224307.1 | - | 5, 6 | 398 | |||
KJ767789.1 | - | 6, 7 | 549 | |||
KJ767790.1 | - | 3, 4 | 167 |