Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190996.1 | NP_001177925.1 | 3 | 257 | ||
NM_006409.3 | NP_006400.2 | 3 | 259 | |||
GenBank mRNA | AB209059.1 | - | 3 | 192 | ||
AK223015.1 | - | 2 | 83 | |||
BC039594.1 | - | 3 | 222 | 6018804 | ||
BC047889.1 | - | 3 | 192 | 6473050 | ||
BC054027.1 | - | 3 | 191 | 6514889 | ||
BM924483.1 | - | 245 | ||||
DA089818.1 | - | 3 | 259 | |||
EU668326.1 | - | 3 | 245 | |||
GQ472234.1 | - | 2 | 83 | |||
Y08999.1 | - | 2 | 131 |