Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 2 | 268 | ||
NM_001098521.1 | NP_001091991.1 | 2 | 196 | |||
NM_001098522.1 | NP_001091992.1 | 1 | 478 | |||
NM_001098523.1 | NP_001091993.1 | 1 | 194 | |||
NM_006410.4 | NP_006401.3 | 2 | 163 | |||
GenBank mRNA | AF039103.1 | - | 1 | 109 | ||
AF092095.1 | - | 1 | 195 | |||
AI193677.1 | - | 1 | 478 | |||
AK222969.1 | - | 1 | 152 | |||
AK223010.1 | - | 1 | 186 | |||
AK292092.1 | - | 2 | 172 | |||
AW963626.1 | - | 1 | 113 | |||
BC002439.2 | - | 2 | 209 | 3346984 | ||
BC015358.2 | - | 1 | 174 | 3455757 | ||
BP382529.1 | - | 2 | 268 | |||
CR456821.1 | - | 1 | 97 | |||
DB450790.1 | - | 2 | 195 | |||
U69161.1 | - | 1 | 195 |