Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 6 | 1071 | ||
NM_001098521.1 | NP_001091991.1 | 6 | 999 | |||
NM_001098522.1 | NP_001091992.1 | 5 | 1281 | |||
NM_006410.4 | NP_006401.3 | 6 | 966 | |||
GenBank mRNA | AF039103.1 | - | 5 | 909 | ||
AK130473.1 | - | 1 | 396 | |||
AK222969.1 | - | 5 | 955 | |||
BC002439.2 | - | 6 | 1012 | 3346984 | ||
BC015358.2 | - | 5 | 977 | 3455757 | ||
U69161.1 | - | 5 | 998 |