Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006413.4 | NP_006404.1 | 11 | 953 | ||
XM_017015477.1 | XP_016870966.1 | 12 | 1206 | |||
GenBank mRNA | AL833628.1 | - | 11 | 938 | ||
BC006991.1 | - | 12 | 1212 | 3827681 |