Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097590.2 | NP_001091059.1 | 3 | 1257 | ||
NM_001265601.1 | NP_001252530.1 | 2 | 1495 | |||
NM_006414.4 | NP_006405.2 | 2 | 1138 | |||
NM_183005.4 | NP_892117.1 | 3 | 1614 | |||
XM_006717363.1 | XP_006717426.1 | 3 | 1428 | |||
XM_006717364.3 | XP_006717427.1 | 2 | 1050 | |||
XM_011519293.1 | XP_011517595.1 | 2 | 1293 | |||
XM_017015479.1 | XP_016870968.1 | 3 | 1746 | |||
XM_017015480.1 | XP_016870969.1 | 3 | 1204 | |||
XM_017015481.1 | XP_016870970.1 | 2 | 1228 | |||
GenBank mRNA | AK057006.1 | - | 3 | 1412 | ||
AK223418.1 | - | 2 | 1115 | |||
BC029494.1 | - | 3 | 1395 | 5271044 | ||
U77664.1 | - | 2 | 934 |