Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006419.2 | NP_006410.1 | 5 | 1026 | ||
XM_006714063.3 | XP_006714126.1 | 4 | 1019 | |||
GenBank mRNA | AF044197.1 | - | 5 | 1042 | ||
BC012589.1 | - | 4 | 998 | 4246053 |