Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006461.3 | NP_006452.3 | 3 | 1070 | ||
GenBank mRNA | AF063308.1 | - | 3 | 1047 | ||
AF345347.2 | - | 3 | 1311 | |||
AF399910.1 | - | 3 | 1049 | |||
AK025725.1 | - | 3 | 1042 | |||
AK292398.1 | - | 3 | 1061 | |||
AL137585.1 | - | 1 | 481 | |||
BC000322.2 | - | 3 | 1030 | 2821784 |