Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313972.1 | NP_001300901.1 | 7 | 1748 | ||
NM_006472.5 | NP_006463.3 | 8 | 2117 | |||
XM_017000085.1 | XP_016855574.1 | 7 | 2195 | |||
GenBank mRNA | AK301203.1 | - | 4 | 1237 | ||
BC024305.1 | - | 8 | 1647 | |||
BF446648.1 | - | 1 | 95 | |||
BX404012.2 | - | 1 | 247 | |||
BX537824.1 | - | 5 | 1499 | |||
S73591.1 | - | 8 | 1973 |