Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006473.3 | NP_006464.1 | 6 | 677 | ||
XM_005273714.1 | XP_005273771.1 | 6 | 542 | |||
GenBank mRNA | AF069735.1 | - | 5 | 476 | ||
AK289499.1 | - | 6 | 587 | |||
AK310054.1 | - | 6 | 578 | |||
AK314330.1 | - | 6 | 563 | |||
BC008785.2 | - | 6 | 577 |