Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278730.1 | NP_001265659.1 | 6 | 1696 | ||
NM_006478.4 | NP_006469.2 | 6 | 2405 | |||
NM_152236.2 | NP_689422.1 | 6 | 2410 | |||
XM_011529824.1 | XP_011528126.1 | 7 | 2529 | |||
XM_011529825.1 | XP_011528127.1 | 6 | 2338 | |||
XM_017028533.1 | XP_016884022.1 | 6 | 3055 | |||
GenBank mRNA | AK093386.1 | - | 6 | 2347 | ||
AK122829.1 | - | 5 | 3247 | |||
AK223602.1 | - | 6 | 2349 | |||
AK225484.1 | - | 6 | 1668 | |||
BC001782.1 | - | 6 | 2345 | 3355168 | ||
BC007624.1 | - | 6 | 2345 | 3357387 | ||
BC011047.1 | - | 6 | 2348 | 4178571 |