Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006480.4 | NP_006471.2 | 13 | 1544 | ||
XM_005265794.4 | XP_005265851.1 | 13 | 1714 | |||
XM_005265795.4 | XP_005265852.1 | 13 | 1719 | |||
GenBank mRNA | AF037195.1 | - | 9 | 976 | ||
AK098150.1 | - | 6 | 1469 | |||
AK123382.1 | - | 9 | 888 | |||
AY987041.1 | - | 9 | 906 | |||
BC014094.2 | - | 13 | 1514 | 4547415 |