Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008390.1 | NP_001008391.1 | 4 | 635 | ||
NM_001195308.1 | NP_001182237.1 | 4 | 616 | |||
NM_003663.3 | NP_003654.3 | 3 | 533 | |||
XM_017007355.1 | XP_016862844.1 | 3 | 441 | |||
GenBank mRNA | AJ000258.1 | - | 3 | 319 | ||
AL117392.1 | - | 3 | 479 | |||
BC052980.1 | - | 3 | 435 | 6012191 | ||
CR456854.1 | - | 1 | 123 |